EMAIL THIS PAGE TO A FRIEND
Arthritis and rheumatism
Ischemic reaction secondary to corticosteroid injection of the joint.
PMID 23606212
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1950000 Triamcinolone hexacetonide, European Pharmacopoeia (EP) Reference Standard
C30H41FO7