EMAIL THIS PAGE TO A FRIEND
Transboundary and Emerging Diseases
Polymorphism at the ITS and NTS Loci of Perkinsus marinus isolated from cultivated oyster Crassostrea corteziensis in Nayarit, Mexico and phylogentic relationship to P. marinus along the Atlantic Coast.
PMID 23607743