EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Treatment-emergent effects may predict benefit from endocrine therapy.
PMID 23610120