EMAIL THIS PAGE TO A FRIEND
Mikrobiologiia
Screening and identification of newly isolated cellulose-degrading bacteria from the gut of xylophagous termite Microcerotermes diversus (Silvestri).
PMID 23610931
Abstract
The aim of the present study was to isolate and characterize the cellulose-degrading bacteria from the gut of the local termite, Microcerotermes diversus (Silvestri), inhabiting the Khuzestan province of Iran. The microorganisms capable of growing in the liquid medium containing cellulose as the only source of carbon were isolated and their cellulolytic activity on CMC-containing media was confirmed by the congo red clearing zone assay. The isolates were identified based on biochemical characteristics and the phylogenetic analysis of 16S rRNA gene fragments. The results of the present study show that three cellulose-degrading bacteria isolated from local termite guts belonged to the genera Acinetobacter, Pseudomonas and Staphylococcus and four cellulose-degrading bacteria belonged to Enterobacteriaceae and Bacillaceae families. Several isolates recovered from separate termite Microcerotermes diversus samples closely clustered in phylogenetic trees indicating high similarity and the abundance of particular cellulolytic strains. Bacillus B5B and Acinetobacter L9B hydrolyzed cellulose faster than the other isolates (with CMCase activity of 1.47 U/mL and 1.22 U/mL, respectively). The stability of CMCase produced by Bacillus B5B over a broad range of pH and high temperature indicated that the enzyme may be of great commercial value.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8002
α-Cellulose, powder
C6429
α-Cellulose, BioReagent, suitable for insect cell culture
11363
Avicel® PH-101, tested according to Ph.Eur.
11365
Avicel® PH-101, ~50 μm particle size
C9481
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
C5678
Carboxymethylcellulose sodium salt, low viscosity
C4888
Carboxymethylcellulose sodium salt, Medium viscosity
C5013
Carboxymethylcellulose sodium salt, High viscosity
C1184 Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
22178 Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
C2605
Cellulase from Aspergillus sp., aqueous solution
C2730 Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
C8546
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
C0615
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
C1794 Cellulase from Trichoderma sp., BioReagent, plant cell culture tested, 3-10 units/mg solid
C9499
Cellulase, thermostable from Clostridium thermocellum, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
C6288
Cellulose, fibers, (medium)
22182
Cellulose, acid washed, from spruce, for column chromatography
22184
Cellulose, acid washed, powder, for column chromatography
22183
Cellulose, powder, for column chromatography
22197
Cellulose, DS-0, powder, for thin layer chromatography
09906
Cellulose, DFS-0, microcrystalline, for thin layer chromatography
435244
Cellulose, colloidal, microcrystalline
310697
Cellulose, microcrystalline, powder, 20 μm
435236
Cellulose, microcrystalline, powder
C6277
Congo Red, certified by the Biological Stain Commission, BioXtra
C32H22N6Na2O6S2
75768
Congo Red, analytical standard
C32H22N6Na2O6S2
C6767
Congo Red, Dye content ≥35 %
C32H22N6Na2O6S2
S3504
Sigmacell Cellulose, Type 20, 20 μm
S5504
Sigmacell Cellulose, Type 50, 50 μm