EMAIL THIS PAGE TO A FRIEND
Klinichna khirurhiia
[Application of combined antibacterial preparations in the treatment of surgical intra-abdominal infections].
PMID 23610943
Abstract
Experience of application of combined preparation Ciprolet A (ciprofloxacine 500 mg, tinidazol 600 mg) in 27 patients after performed reconstructive operations for surgical diseases of abdominal organs as well as in 50 patients in destructive forms of an acute cholecystitis was summarized. Possibility of the preparation application in ambulatory conditions constitutes its advantage.