EMAIL THIS PAGE TO A FRIEND
Acta Poloniae Pharmaceutica
Formulation, in vitro drug release study and anticancer activity of 5-fluorouracil loaded gellan gum microbeads.
PMID 23610967
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223506
Calcium chloride dihydrate, ACS reagent, ≥99%
CaCl2 · 2H2O
C3881
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
CaCl2 · 2H2O
31307
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
CaCl2 · 2H2O
12022
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
CaCl2 · 2H2O
200646
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
200689
Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
200697
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
433837
Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
247499
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
200662
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 49% ethoxyl
200654
Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
E8003
Ethyl cellulose, extent of labeling: 48-49.5% ethoxy content
46070
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
46080
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
28244
Ethyl cellulose, tested according to Ph.Eur., viscosity 10 mPa.s 
Z0251
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
O4SZn · 7H2O
Z1001
Zinc sulfate heptahydrate, plant cell culture tested
O4SZn · 7H2O
Z0635
Zinc sulfate heptahydrate, meets USP testing specifications
O4SZn · 7H2O
204986
Zinc sulfate heptahydrate, 99.995% trace metals basis
O4SZn · 7H2O
Z4750
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
O4SZn · 7H2O
221376
Zinc sulfate heptahydrate, ACS reagent, 99%
O4SZn · 7H2O
31665
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
O4SZn · 7H2O
14455
Zinc sulfate heptahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-103.0%
O4SZn · 7H2O
96497
Zinc sulfate heptahydrate, tested according to Ph.Eur.
O4SZn · 7H2O
83265
Zinc sulfate solution, BioUltra, for molecular biology, 2.0 M in H2O
O4SZn
Z2876
Zinc sulfate solution, 0.3 N
O4SZn