EMAIL THIS PAGE TO A FRIEND
Genes to cells : devoted to molecular & cellular mechanisms
Interaction of Necl-4/CADM4 with ErbB3 and integrin α6 β4 and inhibition of ErbB2/ErbB3 signaling and hemidesmosome disassembly.
PMID 23611113
Abstract
Nectin-like molecule 4 (Necl-4)/CADM4, a transmembrane cell-cell adhesion molecule with three Ig-like domains, was shown to serve as a tumor suppressor, but its mode of action has not been elucidated. In this study, we showed that Necl-4 interacted in cis with ErbB3 through their extracellular regions, recruited PTPN13 and inhibited the heregulin-induced activation of the ErbB2/ErbB3 signaling. In addition, we extended our previous finding that Necl-4 interacts in cis with integrin α6 β4 through their extracellular regions and found that Necl-4 inhibited the phorbol ester-induced disassembly of hemidesmosomes. These results indicate that Necl-4 serves as a tumor suppressor by inhibiting the ErbB2/ErbB3 signaling and hemidesmosome disassembly.