EMAIL THIS PAGE TO A FRIEND
The Journal of oral implantology
Treatment of congenitally missing maxillary lateral incisors: an interdisciplinary approach.
PMID 23611678
Abstract
Dentists have several treatment options when restoring patients with congenitally missing maxillary lateral incisors. Endosseous implant therapy combined with orthodontics can achieve predictable outcomes while meeting patient expectations. This case report exhibits an interdisciplinary approach involving orthodontics, implant surgery, and prosthodontics to resolve this common dental concern.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
587583
Zinc phosphate, 99.998% trace metals basis
O8P2Zn3