EMAIL THIS PAGE TO A FRIEND
The Journal of oral implantology
OsteoGen crystals coated with a biomimetic nano-crystalline fluorapatite surface technology to promote osteoblast cell differentiation, migration and proliferation.
PMID 23611679