EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Determination of doxycycline in chicken fat by liquid chromatography with UV detection and liquid chromatography-tandem mass spectrometry.
PMID 23611844
Abstract
A sensitive analytical method for determination of doxycycline (DC) residues in chicken fat/fat and skin was developed. The extraction, in the presence of the internal standard (IS) minocycline (MINO), was carried out using solution of oxalic acid (pH 4.0) and ethyl acetate. The samples were cleaned up by solid phase extraction (SPE) procedure using, at first carboxylic acid and then polymeric Strata X cartridges. Chromatographic separation of DC by LC-UV was achieved on a Luna C8 analytical column and for LC-MS/MS analysis Luna C18 column was used. The presented procedures were evaluated according to the Commission Decision 2002/657/EC. Specificity, decision limit (CCα), detection capacity (CCβ), recovery (absolute and relative), precision (repeatability and reproducibility) were determined during validation process. The limit of detection (LOD) was 10μg/kg for LC-UV and 1μg/kg for LC-MS/MS method. The limit of quantitation (LOQ) was 15 and 2μg/kg for LC-UV and LC-MS/MS, respectively. The absolute recovery for the LC-UV and relative recovery for the LC-MS/MS method at 300μg/kg concentration level were 79%; 101% for fat and 82%; 99% for fat and skin, respectively. The developed liquid chromatography with ultraviolet detection (LC-UV) and liquid chromatography tandem mass spectrometry (LC-MS/MS) methods have been applied to quantitative determination of doxycycline (DC) in samples of chicken fat tissue obtained from animals treated with DC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3072 Doxycycline Hydrochloride, Ready Made Solution
C22H24N2O8 · HCl
D1822
Doxycycline monohydrate
C22H24N2O8 · H2O