EMAIL THIS PAGE TO A FRIEND
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova
[Current classifications of dystonies, treatment strategy].
PMID 23612417
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5311
Biperiden hydrochloride, ≥98% (HPLC), powder
C21H30ClNO