EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Smoke-derived karrikin perception by the α/β-hydrolase KAI2 from Arabidopsis.
PMID 23613584
Abstract
Genetic studies in Arabidopsis implicate an α/β-hydrolase, KARRIKIN-INSENSITIVE 2 (KAI2) as a receptor for karrikins, germination-promoting butenolide small molecules found in the smoke of burned plants. However, direct biochemical evidence for the interaction between KAI2 and karrikin and for the mechanism of downstream signaling by a KAI2-karrikin complex remain elusive. We report crystallographic analyses and ligand-binding experiments for KAI2 recognition of karrikins. The karrikin-1 (KAR1) ligand sits in the opening to the active site abutting a helical domain insert but distal from the canonical catalytic triad (Ser95-His246-Asp217) of α/β-hydrolases, consistent with the lack of detectable hydrolytic activity by purified KAI2. The closest approach of KAR1 to Ser95-His246-Asp217 is 3.8 Å from His246. Six aromatic side chains, including His246, encapsulate KAR1 through geometrically defined aromatic-aromatic interactions. KAR1 binding induces a conformational change in KAI2 at the active site entrance. A crevice of hydrophobic residues linking the polar edge of KAR1 and the helical domain insert suggests that KAI2-KAR1 creates a contiguous interface for binding signaling partners in a ligand-dependent manner.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
333131
N,N′-Bis(2-aminoethyl)-1,3-propanediamine, 97%
C7H20N4