EMAIL THIS PAGE TO A FRIEND
Molecular plant-microbe interactions : MPMI
Nicotiana benthamiana calreticulin 3a is required for the ethylene-mediated production of phytoalexins and disease resistance against oomycete pathogen Phytophthora infestans.
PMID 23617417
Abstract
Mature Nicotiana benthamiana shows strong resistance to the potato late blight pathogen Phytophthora infestans. By screening using virus-induced random gene silencing, we isolated a gene for plant-specific calreticulin NbCRT3a as a required gene for resistance of N. benthamiana against P. infestans. NbCRT3a encodes an endoplasmic reticulum quality-control (ERQC) chaperone for the maturation of glycoproteins, including glycosylated cell-surface receptors. NbCRT3a-silenced plants showed no detectable growth defects but resistance to P. infestans was significantly compromised. Defense responses induced by the treatment with INF1 (a secretory protein of P. infestans), such as production of reactive oxygen species and accumulation of phytoalexins, were suppressed in NbCRT3a-silenced N. benthamiana. Expression of an ethylene-regulated gene for phytoalexin biosynthesis, NbEAS, was reduced in NbCRT3a-silenced plants, whereas the expression of salicylic acid-regulated NbPR-1a was not affected. Consistently, induction of ethylene production by INF1 was suppressed in NbCRT3a-silenced plants. Resistance reactions induced by a hyphal wall components elicitor prepared from P. infestans were also impaired in NbCRT3a-silenced plants. However, cell death induced by active mitogen-activated protein kinase kinase (NbMEK2(DD)) was not affected by the silencing of NbCRT3a. Thus, NbCRT3a is required for the initiation of resistance reactions of N. benthamiana in response to elicitor molecules derived from P. infestans.