EMAIL THIS PAGE TO A FRIEND
Postepy higieny i medycyny doswiadczalnej (Online)
In vivo anti-inflammatory activity of lipoic acid derivatives in mice.
PMID 23619233
Abstract
In mammals lipoic acid (LA) and its reduced form dihydrolipoic acid (DHLA) function as cofactors for multienzymatic complexes catalyzing the decarboxylation of α-ketoacids. Moreover, LA is used as a drug in a variety of diseases including inflammatory diseases. The aim of the study was to examine anti-inflammatory properties of LA metabolites. The present paper reports the chemical synthesis of 2,4-bismethylthio-butanoic acid (BMTBA) and tetranor-dihydrolipoic acid (tetranor-DHLA). BMTBA is one of the biotransformation products of LA, while tetranor-DHLA is an analogue of DHLA. Structural identity of these compounds was confirmed by 1H NMR. These compounds were assessed for their anti-inflammatory activity in mice. For this purpose, the zymosan-induced peritonitis and the carrageenan-induced hind paw edema animal models were applied. The obtained results indicated that the early vascular permeability measured at 30 min of zymosan-induced peritonitis was significantly inhibited in groups receiving BMTBA (10, 30, 50 mg/kg). The early infiltration of neutrophils measured at 4 hours of zymosan-induced peritonitis was inhibited in the group receiving BMTBA (50 mg/kg) and tetranor-DHLA (50 mg/kg). The results indicated that the increase in paw edema was significantly inhibited in the groups receiving BMTBA (50, 100 mg/kg) and tetranor-DHLA (30, 50 mg/kg). In summary, the present studies clearly demonstrated that both BMTBA and tetranor-DHLA were able to act as anti-inflammatory agents. This is the first study examining in vivo the anti-inflammatory properties of LA metabolites.