EMAIL THIS PAGE TO A FRIEND
Endocrine
Is there a role for combination therapy for osteoporosis?
PMID 23619961
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0650 Risedronate sodium, ≥97% (HPLC)
C7H10NO7P2 · Na