EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Determination of a dantrolene metabolite, 5-(p-nitrophenyl)-2-furoic acid, in plasma and urine by high-performance liquid chromatography.
PMID 2361999
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
428558
5-(4-Nitrophenyl)-2-furoic acid, 96%
C11H7NO5