EMAIL THIS PAGE TO A FRIEND
The West Indian medical journal
Adverse reaction of topical etofenamate: petechial eruption.
PMID 23620980
Abstract
Etofenamate is a non-steroidal anti-inflammatory drug (NSAID). Clinical findings caused by etofenamate are uncommon. Allergic contact dermatitis is the most common cutaneous reaction reported. But petechial eruption due to etofenamate had not been reported yet. This report concerns an 11-year old male with petechial eruption after application of topical etofenamate. Physicians need to be aware that patients can develop an asymptomatic purpuric eruption when etofenamate is ordered.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2499900 Etofenamate, European Pharmacopoeia (EP) Reference Standard
C18H18F3NO4