EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Effect of paracetamol use on the modification of the development of asthma by reactive oxygen species genes.
PMID 23622008
Abstract
Recent studies have identified an increase in the prevalence of asthma associated with paracetamol use. To identify the relationship among asthma, biomarkers, genes, and paracetamol use in preschool children. We undertook a population-based, cross-sectional survey of 933 preschool children. Asthma status was classified according to medical history and asthmatic symptoms. History of paracetamol use in infancy was recorded. Impulse oscillometry, blood tests for eosinophils and total IgE, and genotyping of NAT2, Nrf2, and GSTP1 polymorphisms by TaqMan assay were conducted. Paracetamol use in infancy was associated with an increased risk of treatment for asthma within the previous 12 months. Paracetamol use together with a family history of asthma increased the risk of asthma diagnosis ever, current asthma, and treatment for asthma within the previous 12 months. Gene polymorphisms in NAT2 (rs4271002), Nrf2 (rd6726395), and GSTP1 (rd1695) increased the risk of treatment for asthma within the last 12 months. Eosinophils were significantly elevated in the group with paracetamol use and a family history of asthma; however, the serum total IgE level and IOS did not show any significant difference. Paracetamol use in infancy was significantly associated with increased risk of asthma. The association is more significant in genetically susceptible children, related to antioxidant genes, and the effect may be mediated by eosinophilic inflammation.