EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Acute generalized exanthematic pustulosis due to ibuprofen.
PMID 23622012