EMAIL THIS PAGE TO A FRIEND
Journal of dermatological science
Mild phenotype of epidermolytic hyperkeratosis mimicking ichthyosis bullosa of Siemens is related to specific mutation in 2B domain of KRT1.
PMID 23623204