EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
The interaction of adenosine and morphine on pentylenetetrazole-induced seizure threshold in mice.
PMID 23624288
Abstract
Adenosine agonists or low doses of morphine exert anti-convulsant effects in different models of seizures. On the other hand, a tight interaction has been reported between morphine and adenosine in various paradigms. This study investigated the effect of the interaction of adenosine and morphine on seizure susceptibility in the intravenous mouse model of pentylenetetrazole (PTZ)-induced clonic seizures. The researchers used acute systemic administration of morphine, N(6)-cyclohexyladenosine (CHA) (a selective A1 receptor agonist), naltrexone (an opioid receptor antagonist) and 8-Cyclopentyl-1,3-dimethylxanthine (8-CPT) (a selective A1 receptor antagonist). Acute administration of morphine (0.25, 0.5 and 1 mg/kg) or CHA (0.25, 0.5, 1, 2 and 4 mg/kg) raised the threshold of seizures induced by PTZ. Non-effective dose of 8-CPT (2 mg/kg) inhibited the anticonvulsant effects of CHA (0.5 and 1 mg/kg). Combination of sub-effective doses of morphine (0.125 mg/kg) and CHA (0.125 mg/kg) increased clonic seizure latency showing the additive effect of morphine and CHA. The enhanced latency induced by combination of low doses of morphine and CHA completely reversed by 8-CPT (2 mg/kg) or naltrexone (1 mg/kg). Moreover, 8-CPT (2 mg/kg) inhibited anticonvulsant effects of morphine (0.25 and 0.5 mg/kg) and naltrexone (1 mg/kg) inhibited anticonvulsant effects of CHA (0.25, 0.5 and 1 mg/kg). Combination of low doses of 8-CPT (1 mg/kg) and naltrexone (0.5 mg/kg) inhibited the anticonvulsant effect of CHA (0.5 and 1 mg/kg). In conclusion, adenosine and morphine exhibit an additive effect on the enhancement of the pentylenetetrazole-induced seizure threshold in mice, probably through A1 or μ receptors.