EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
PGI₂ signaling inhibits antigen uptake and increases migration of immature dendritic cells.
PMID 23625201
Abstract
PGI₂ signaling through IP inhibits allergen-induced inflammatory responses in mice. We reported previously that PGI₂ analogs decreased proinflammatory cytokine and chemokine production by mature BMDCs. However, whether PGI₂ modulates the function of immature DCs has not been investigated. We hypothesized that PGI2 negatively regulates immature DC function and investigated the effect of PGI2 analogs on immature BMDC antigen uptake and migration in vitro and in vivo. Immature BMDCs were obtained from WT and IPKO mice, both on a C57BL/6 background. The PGI2 analog cicaprost decreased FITC-OVA uptake by immature BMDCs. In addition, cicaprost increased immature BMDC podosome dissolution, pro-MMP-9 production, cell surface CCR7 expression, and chemotactic migration toward CCL19 and CCL21, as well as chemokinesis, in an IP-specific fashion. These in vitro results suggested that cicaprost promotes migration of immature DCs from mucosal surface to draining LNs. This concept was supported by the finding that migration of immature GFP⁺ BMDCs to draining LNs was enhanced by pretreatment with cicaprost. Further, migration of immature lung DCs labeled with PKH26 was enhanced by intranasal cicaprost administration. Our results suggest PGI2-IP signaling increases immature DC migration to the draining LNs and may represent a novel mechanism by which this eicosanoid inhibits immune responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6188
Prostaglandin I2 sodium salt, ≥96% (HPLC), synthetic, powder
C20H31NaO5