EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Hansschlegelia beijingensis sp. nov., an aerobic, pink-pigmented, facultatively methylotrophic bacterium isolated from watermelon rhizosphere soil.
PMID 23625265
Abstract
A novel Gram-stain-negative, aerobic, rod-shaped strain designated PG04(T) was isolated from the rhizosphere of watermelon plants cultivated in Beijing, China. A polyphasic taxonomic study was performed on the new isolate. On the basis of 16S rRNA gene sequence similarity studies, isolate PG04(T) belonged clearly to the genus Hansschlegelia and was most closely related to Hansschlegelia zhihuaiae (97.3 % similarity to the type strain). The predominant respiratory quinone was ubiquinone 10 (Q-10) and the polar lipid profile was composed of the major lipids diphosphatidylglycerol, phosphatidylglycerol, phosphatidylethanolamine and phosphatidylcholine. The major fatty acids were C18 : 1ω7c (41.3 %), C19 : 0 cyclo ω8c (30.6 %) and C16 : 0 (19.1 %). The G+C content of the DNA was about 64.4 mol%. DNA-DNA hybridization experiments showed 34.4 % relatedness between strain PG04(T) and H. zhihuaiae DSM 18984(T). The results of physiological and biochemical tests and differences in fatty acid profiles allowed clear phenotypic differentiation of strain PG04(T) from the most closely related species in the genus, H. zhihuaiae. Strain PG04(T) therefore represents a novel species within the genus Hansschlegelia, for which the name Hansschlegelia beijingensis sp. nov. is proposed, with the type strain PG04(T) ( = DSM 25481(T) = ACCC 05759(T)).