EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Study on chemical constituents of root bark of Discocleidion rufescens].
PMID 23627092
Abstract
To study the chemical constituents from root bark of Discocleidion rufescens. Column chromatography and spectral analysis were used to isolate and identify the constituents. Ten compounds were obtained and identified from root bark of Discocleidion rufescens including beta-sitosterol (I), scopoletin (II), daucosterol (III), ricinine (IV), chrysophanol (V), aphyscion (VI), taraxerol (VII), pigenin (VIII), luteolin (IX), gallic acid (X). Ten compounds are isolated for the first time from root bark of Discocleidion rufescens. The monomer of Compounds II is isolated from this genus for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75934
3β-Taraxerol, analytical standard
C30H50O