EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents from cell suspension cultures of Cudrania tricuspidata].
PMID 23627169
Abstract
Ten compounds were isolated and purified from cell suspension cultures of Cudrania tricuspidata with silica-gel column chromatography, semi-preparative HPLC and Sephadex LH-20. On the base of their physicochemical properties and spectral data, their structures were identified as 1, 3, 5-trihydroxy4-(3, 3-dimethylallyl) xanthone (1), wighteone (2), 6-prenylapigenin (3), licoflavone C(4), cudraflavanone C(5), erythrivarone A (6), derrone (7), carthamidin (8), genistein (9) and aromadendrin (10). Among them, compounds 2-10 were flavonoids, and compound 1 was a xanthone which was isolated from the plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92204
Dihydrokaempferol, analytical standard
C15H12O6
X600
Xanthone, 97%
C13H8O2