EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Study on optimization of induction system of test-tube tuberous roots from leaves of Rehmannia glutinosa].
PMID 23627185
Abstract
To study the effect of sucrose and plant growth substances of different concentrations on the induction of test-tube tuberous roots of Rehmannia glutinosa, in order to establish an efficient system for the induction of test-tube tuberous roots from leaves of R. glutinosa. Leaves from test-tube seedlings of 85-5 R. glutinosa were used as explants. After rooting induction, they were transferred to medium with orthogonal design for inducing test-tube tuberous roots of R. glutinosa. NAA played a significant role in induction of test-tube tuberous roots of R. glutinosa, followed by sucrose and 6-BA. With leaves from test-tube seedlings as the explants, the optimal medium for inducing test-tube tuberous roots of R. glutinosa was MS + BA 3.0 mg x L(-1) + NAA 0.1 mg x L(-1) + sucrose 7%. The study provides an efficient induction system for studies on artificial seeds and secondary metabolism with test-tube tuberous roots of R. glutinosa.