EMAIL THIS PAGE TO A FRIEND
Cancer research
BMP-6 in renal cell carcinoma promotes tumor proliferation through IL-10-dependent M2 polarization of tumor-associated macrophages.
PMID 23633487
Abstract
Dysregulated bone morphogenetic proteins (BMP) may contribute to the development and progression of renal cell carcinoma (RCC). Herein, we report that BMP-6 promotes the growth of RCC by interleukin (IL)-10-mediated M2 polarization of tumor-associated macrophages (TAM). BMP-6-mediated IL-10 expression in macrophages required Smad5 and STAT3. In human RCC specimens, the three-marker signature BMP-6/IL-10/CD68 was associated with a poor prognosis. Furthermore, patients with elevated IL-10 serum levels had worse outcome after surgery. Together, our results suggest that BMP-6/macrophage/IL-10 regulates M2 polarization of TAMs in RCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3072 Doxycycline Hydrochloride, Ready Made Solution
C22H24N2O8 · HCl
D1822
Doxycycline monohydrate
C22H24N2O8 · H2O