EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
Infectious canine hepatitis associated with prednisone treatment.
PMID 23633720
Abstract
An 11-week-old, female Alaskan husky dog housed outdoors in the Yukon, Canada, was diagnosed with infectious canine hepatitis. The predisposing factors in this puppy for such a rare disease included inappropriate vaccination program, potential contact with endemic wildlife, and immunosuppression due to prednisone treatment.