EMAIL THIS PAGE TO A FRIEND
Blood
Malaria during pregnancy with parasite sequestration in the villous chamber.
PMID 23634437
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1159 Clindamycin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C18H33ClN2O5S · HCl · H2O