EMAIL THIS PAGE TO A FRIEND
Middle East journal of anaesthesiology
Combined use of metoclopramide and dexamethasone as a prophylactic antiemetic in elective cesarean section under spinal anesthesia.
PMID 23634564
Abstract
Nausea and vomiting during regional anesthesia for cesarean section still remain a major problem. We compared the efficacy of dexamethasone plus metoclopramide with dexamethasone alone for preventing nausea and vomiting during and after spinal anesthesia for cesarean section in parturients. The study was performed in 72 full-term parturient women of ASA I & II (American Society of Anesthesiology Grade I & II), aged between 19 and 37 years with uncomplicated pregnancies. The group I (n = 36) received 8 mg of dexamethasone intravenously immediately when the surgery started, while group II (n = 36) received 8 mg of dexamethasone plus 10 mg of metoclopramide. The type and number of episodes of nausea and emesis were recorded, as well as any other adverse effects. During the intraoperative period, a complete response (no emesis, no rescue) was noticed in 83% of patients in Group I and in 86% of patients in Group II. The incidence of nausea during both intra and postoperative periods was not different between the two groups. Metoclopramide was associated with impaired taste and smell and hot flushes. 10 mg of metoclopramide did not improve the incidence of emetic symptoms in patients undergoing cesarean section when combined with 8 mg of dexamethasone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32473
Metoclopramide, VETRANAL®, analytical standard
C14H22ClN3O2