EMAIL THIS PAGE TO A FRIEND
Middle East journal of anaesthesiology
Naloxone versus metoclopramide for the treatment of established postoperative nausea and vomiting in patients following general anesthesia with fentanyl supplementation--pilot study.
PMID 23634567