EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The N-terminal region of two-pore channel 1 regulates trafficking and activation by NAADP.
PMID 23634879
Abstract
TPCs (two-pore channels) are NAADP (nicotinic acid-adenine dinucleotide phosphate)-sensitive Ca2+-permeable ion channels expressed on acidic organelles. In the present study we show that deletion of the N-terminal region redirects TPC1 to the ER (endoplasmic reticulum). The introduction of fluorophores at the N-terminus of TPC1 does not affect its subcellular location, but does reversibly abolish NAADP sensitivity. Our results reveal a dual role for the N-terminus in localization and function of TPC1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5655
Nicotinic acid adenine dinucleotide phosphate sodium salt, ≥92.5%
C21H27N6O18P3 · xNa+