EMAIL THIS PAGE TO A FRIEND
Irish journal of medical science
Contamination of blood pressure cuffs by methicillin-resistant Staphylococcus aureus and preventive measures.
PMID 23639972
Abstract
Although blood pressure cuffs are commonly used and shared in medical facilities, their routine disinfection is performed infrequently. We investigated the contamination of blood pressure cuffs by methicillin-resistant Staphylococcus aureus (MRSA). The MRSA level on the inner side (the surface in contact with patients' skin) of blood pressure cuffs used in the wards and outpatient clinics of a university hospital (733 beds) was determined using the gauze and swab wiping methods. Using the gauze wiping method (n = 35), the MRSA contamination rate was 31.4 %, and the MRSA contamination level was 1,702.6 ± 9,996.1 (0-58, 320) colony-forming units (cfu)/cuff. No MRSA was detected on blood pressure cuffs after washing (n = 30) or wiping with 80 vol% ethanol (n = 18). Blood pressure cuffs are frequently contaminated by MRSA.