EMAIL THIS PAGE TO A FRIEND
Pathology oncology research : POR
Synergic insulin sensitizing effect of rimonabant and BGP-15 in Zucker-obese rats.
PMID 23640247
Abstract
Abdominal obesity is referred for as a common pathogenic root of multiple risk factors, which include insulin resistance, dyslipidemia, hypertension, and a pro-atherogenic and pro-inflammatory state. Irrespective of its psychiatric side effects, rimonabant through blocking cannabinoid-1 receptor (CB1R) induces an increase in whole body insulin sensitivity. The aim of this work was to study the effect of selected doses of another insulin sensitizer compound BGP-15, and rimonabant on insulin resistance in Zucker obese rats with a promise of inducing insulin sensitization together at lower doses than would have been expected by rimonabant alone. We found that BGP-15 potentiates the insulin sensitizing effect of rimonabant. The combination at doses, which do not induce insulin sensitization by themselves, improved insulin signaling. Furthermore our results suggest that capsaicin-induced signal may play a role in insulin sensitizing effect of both molecules. Our data might indicate that a lower dose of rimonabant in the treatment of insulin resistance and type 2 diabetes is sufficient to administer, thus a lower incidence of the unfavorable psychiatric side effects of rimonabant are to be expected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0800 Rimonabant hydrochloride, ≥98% (HPLC)
C22H21Cl3N4O · HCl