EMAIL THIS PAGE TO A FRIEND
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia
Outcome of individualized dialysate sodium concentration for hemodialysis patients.
PMID 23640622
Abstract
To evaluate the individualization of dialysate sodium (Na + ) concentration in hemodialysis (HD), we studied 40 stable chronic HD patients in a single-blind crossover design. They underwent 36 consecutive HD sessions with the dialysate Na + concentration set at 138 mmol/L, followed by 36 sessions of dialysate Na + set to match the patients average pre-HD plasma Na + levels. We multiplied the midweek pre-HD measured Na + by the Donnan coefficient of 0.95 (individualized Na + ). Pre-HD Na + dialysis sodium levels were nearly constant, with no variation between the two phases and a mean of 137.45 ± 2.04 mmol/L. Post-HD serum Na + was significantly higher during the standard phase (139.7 ± 2 mmol/L) than during the individualized phase (137.1 ± 1.6 mmol/L). Also, interdialytic weight gain (IDWG) was significantly more reduced during the individualized phase (3.25 ± 0.56%) than during the standard phase (3.94 ± 0.92%), P <0.001. Episodes of distressing symptoms including headache, muscle cramps and hypotension were significantly less frequent in the individualized phase. The mean of the pre-HD and post-HD systolic and diastolic blood pressures significantly decreased during the individualized phase, and we could reduce the doses of antihypertensive drugs in 10 (33.33%) patients. Individualized dialysate Na + concentration was associated with a decrease in IDWG and dialysis hypotension and related symptoms and better BP control in stable chronic HD patients.