EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Highly site-specific oxygenation of 1-methylhistidine and its analogue with a copper (II)/ascorbate-dependent redox system.
PMID 2364090
Abstract
The reaction of histidine-related materials with copper(II) and ascorbate under physiological conditions has been studied chemically. We discovered that 1-methylimidazole and its analogues, including biological metabolites L-1-methylhistidine and anserine (beta-alanyl-L-1-methylhistidine), exhibited dramatic reactivity with copper(II)/ascorbate. Reaction of copper(II) and ascorbate occurs specifically at the C-2 position of the imidazole ring of L-1-methylhistidine and anserine derivatives with mono-oxygenation to give the 1-methyl-2-imidazolones in good to excellent yields (70-80%). The occurrence of an oxocopper(III) intermediate in the oxidation process of ascorbate is postulated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67520
1-Methyl-L-histidine, ≥98.0% (TLC)
C7H11N3O2