EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Complex N-heterocycle synthesis via iron-catalyzed, direct C-H bond amination.
PMID 23641113
Abstract
The manipulation of traditionally unreactive functional groups is of paramount importance in modern chemical synthesis. We have developed an iron-dipyrrinato catalyst that leverages the reactivity of iron-borne metal-ligand multiple bonds to promote the direct amination of aliphatic C-H bonds. Exposure of organic azides to the iron dipyrrinato catalyst furnishes saturated, cyclic amine products (N-heterocycles) bearing complex core-substitution patterns. This study highlights the development of C-H bond functionalization chemistry for the formation of saturated, cyclic amine products and should find broad application in the context of both pharmaceuticals and natural product synthesis.