EMAIL THIS PAGE TO A FRIEND
International journal for parasitology. Drugs and drug resistance
Identification of plumbagin and sanguinarine as effective chemotherapeutic agents for treatment of schistosomiasis.
PMID 23641325
Abstract
Schistosomiasis, a snail-borne parasitic disease, affects more than 200 million people worldwide. Currently the treatment of schistosomiasis relies on a single therapy of praziquantel, a drug developed over 30 years ago. Thus, there is an urgent need to develop alternative antischistosomal drugs. In the pursuit of novel antischistosomal drugs, we examined the antischistosomal activities of 45 compounds that had been reported to exhibit antimicrobial and/or antiparasitic activities. Two plant-derived compounds, plumbagin and sanguinarine, were found to possess potent antischistosomal activities
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H46805
2-Hydroxy-1,4-naphthoquinone, 97%
C10H6O3
S5890
Sanguinarine chloride hydrate, ≥98% (HPLC)
C20H14ClNO4 · xH2O