EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Cryptosporioptide: a bioactive polyketide produced by an endophytic fungus Cryptosporiopsis sp.
PMID 23642454
Abstract
An antibiotic polyketide, Cryptosporioptide (1) was isolated from the culture extract of the endophytic fungus Cryptosporiopsis sp. The structure of Cryptosporioptide has been established with the help of 1D ((1)H, (13)C), 2D NMR (HSQC, HMBC, COSY, NOESY) techniques and mass spectrometry (FABMS, HRFABMS). The absolute configuration was established by means of electronic circular dichroism (ECD). Cryptosporioptide exhibited both lipoxygenase inhibitory and anti-Bacillus megaterium activities.