EMAIL THIS PAGE TO A FRIEND
Developmental cell
EGF-induced centrosome separation promotes mitotic progression and cell survival.
PMID 23643362
Abstract
Timely and accurate assembly of the mitotic spindle is critical for the faithful segregation of chromosomes, and centrosome separation is a key step in this process. The timing of centrosome separation varies dramatically between cell types; however, the mechanisms responsible for these differences and its significance are unclear. Here, we show that activation of epidermal growth factor receptor (EGFR) signaling determines the timing of centrosome separation. Premature separation of centrosomes decreases the requirement for the major mitotic kinesin Eg5 for spindle assembly, accelerates mitosis, and decreases the rate of chromosome missegregation. Importantly, EGF stimulation impacts upon centrosome separation and mitotic progression to different degrees in different cell lines. Cells with high EGFR levels fail to arrest in mitosis upon Eg5 inhibition. This has important implications for cancer therapy because cells with high centrosomal response to EGF are more susceptible to combinatorial inhibition of EGFR and Eg5.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
164739
(+)-S-Trityl-L-cysteine, 97%
C22H21NO2S
C3290000
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
C3H7NO2S · HCl · H2O
E1257 Epidermal Growth Factor from murine submaxillary gland, lyophilized powder, BioReagent, suitable for cell culture
C7880
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
C3H7NO2S · HCl · H2O
C6852
L-Cysteine hydrochloride monohydrate, from non-animal source, cell culture tested, meets EP, USP testing specifications
C3H7NO2S · HCl · H2O
C5610
L-Cysteine hydrochloride monohydrate, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S · HCl · H2O
30122
L-Cysteine hydrochloride monohydrate, tested according to Ph Eur
C3H7NO2S · HCl · H2O
W779989
L-Cysteine hydrochloride monohydrate, FG, produced by Wacker Chemie AG, Burghausen, Germany, Food, 20 mesh
C3H7NO2S · HCl · H2O
PHR1102
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2S · HCl · H2O
30129
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
778451
L-Cysteine hydrochloride monohydrate, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.5-101.0% (Ph.Eur (2.2.20))
C3H7NO2S · HCl · H2O