EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
A review on zinc and nickel adsorption on natural and modified zeolite, bentonite and vermiculite: examination of process parameters, kinetics and isotherms.
PMID 23644019
Abstract
Adsorption and ion exchange can be effectively employed for the treatment of metal-contaminated wastewater streams. The use of low-cost materials as sorbents increases the competitive advantage of the process. Natural and modified minerals have been extensively employed for the removal of nickel and zinc from water and wastewater. This work critically reviews existing knowledge and research on the uptake of nickel and zinc by natural and modified zeolite, bentonite and vermiculite. It focuses on the examination of different parameters affecting the process, system kinetics and equilibrium conditions. The process parameters under investigation are the initial metal concentration, ionic strength, solution pH, adsorbent type, grain size and concentration, temperature, agitation speed, presence of competing ions in the solution and type of adsorbate. The system's performance is evaluated with respect to the overall metal removal and the adsorption capacity. Furthermore, research works comparing the process kinetics with existing reaction kinetic and diffusion models are reviewed as well as works examining the performance of isotherm models against the experimental equilibrium data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
643653
Aluminosilicate, mesostructured, MCM-41 (hexagonal)
285234 Bentonite
682659 Nanoclay, hydrophilic bentonite
268259
Nickel, foil, thickness 0.125 mm, ≥99.9%
Ni
203904
Nickel, powder, <150 μm, 99.99% trace metals basis
Ni
GF28024657
Nickel, foam, 150x150mm, thickness 1.6mm, bulk density 0.45g/cm3, porosity 95%, 95%
Ni
577995
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Ni
266981
Nickel, powder, <50 μm, 99.7% trace metals basis
Ni
268283
Nickel, powder, <1 μm, 99.8% trace metals basis
Ni
267058
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Ni
267074
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Ni
357553
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Ni
GF04391591
Nickel, mesh, 100x250mm, nominal aperture 0.73mm, wire diameter 0.25mm, 26x26 wires/inch, open area 55%, plain weave mesh, 99%
Ni
357588
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Ni
267007
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Ni
GF66892922
Nickel, wire reel, 100m, diameter 0.5mm, hard, 99.98%
Ni
357626
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ni
GF86176963
Nickel, foil, light tested, 100x100mm, thickness 0.025mm, 99.9%
Ni
357634
Nickel, wire, diam. 0.25 mm, ≥99.9%
Ni
GF84971379
Nickel, mesh, 260x260mm, nominal aperture 0.34mm, thickness 0.025mm, wire diameter 0.041mm, 70 wires/inch, open area 80%, electro-formed mesh, 99.9%
Ni
GF64264181
Nickel, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.98%
Ni
GF45846456
Nickel, wire reel, 1m, diameter 1.0mm, hard, 99.98%
Ni
GF85053988
Nickel, sphere, 500pcs, diameter 3.0mm, precision sphere grade 200, 99%
Ni
GF61615739
Nickel, wire reel, 1m, diameter 1.0mm, as drawn, 99%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF17660594
Nickel, wire reel, 1m, diameter 2.0mm, hard, 99.98%
Ni
GF10370618
Nickel, mesh, 30x30mm, nominal aperture 0.04mm, thickness 0.004mm, wire diameter 0.011mm, 500 wires/inch, open area 60%, electro-formed mesh, 99.9%
Ni
767484
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ni
GF06267652
Nickel, wire reel, 2m, diameter 0.4mm, as drawn, 99.99+%
Ni
GF44869731
Nickel, wire reel, 20m, diameter 0.025mm, as drawn, 99.98%
Ni