EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Study of palladium catalyzation for electroless copper plating on polyimide film.
PMID 23646765
Abstract
In order to form flexible printed circuits through inkjet printing technique, the Pd(ll) catalyst ink was printed on the surface of polyimide film modified with KOH solution and then reduced with NaBH4 solution to extract the Pd(O) catalyst nuclei. The concentration of the Pd(ll) catalyst ink and reduction time showed a significant influence on the microstructure of the Pd(O) catalyst nuclei and the formation of Cu patterns through electroless plating. When reduction time exceeded 1 minute, and as the concentration of the Pd(II) catalyst ink increased above 0.02 M, the catalyst nuclei began aggregation, resulting in Cu patterns with thick and more defects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326666
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Pd
686468
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Pd
203939
Palladium, powder, 99.995% trace metals basis
Pd
464651
Palladium, powder, <75 μm, 99.9% trace metals basis
Pd
203998
Palladium, granular, 99.99% trace metals basis
Pd
267082
Palladium, sponge, 99.9% trace metals basis
Pd
267112
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Pd
373206
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Pd
348643
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Pd
287474
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Pd
348694
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Pd
267120
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Pd
348678
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Pd
411450
Palladium, foil, thickness 0.05 mm, 99.9% trace metals basis
Pd
373192
Palladium, evaporation slug, diam. × L 0.9 cm × 1.2 cm, 99.95% trace metals basis
Pd
346985
Palladium, rod, diam. 6.25 mm, ≥99.9% trace metals basis
Pd
643181
Palladium, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W
Pd
520810
Palladium black, surface area 40-60 m2/g, 99.95% trace metals basis
Pd