EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Synthesis, structure and gas-sensing properties of Pd-functionalized ZnSnO3 rods.
PMID 23646768
Abstract
ZnSnO3 one-dimensional (1D) strutures were synthesized by using an evaporation technique. The morphology, crystal structure, and enhanced sensing properties of the ZnSnO3 structures functionalized with Pd to CO gas at 300 degrees C were investigated. The diameters of the 1D structures ranged from a few hundreds to a few thousands of nanometers and that the lengths were up to a few hundreds of micrometers. The gas sensors fabricated from multiple networked ZnSnO3 rods functionalized with Pd showed the enhanced electrical responses to CO gas. The responses of the rods were improved 10.7, 13.7, 13.4, and 12.5 fold at the CO concentrations of 10, 25, 50, and 100 ppm, respectively. In addition, the mechanism for the enhancement in the gas sensing properties of ZnSnO3 rods by Pd functionalization is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326666
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Pd
686468
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Pd
203939
Palladium, powder, 99.995% trace metals basis
Pd
464651
Palladium, powder, <75 μm, 99.9% trace metals basis
Pd
203998
Palladium, granular, 99.99% trace metals basis
Pd
267082
Palladium, sponge, 99.9% trace metals basis
Pd
267112
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Pd
373206
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Pd
348643
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Pd
287474
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Pd
348694
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Pd
267120
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Pd
348678
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Pd
411450
Palladium, foil, thickness 0.05 mm, 99.9% trace metals basis
Pd
373192
Palladium, evaporation slug, diam. × L 0.9 cm × 1.2 cm, 99.95% trace metals basis
Pd
346985
Palladium, rod, diam. 6.25 mm, ≥99.9% trace metals basis
Pd
643181
Palladium, extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E105CA/W
Pd
520810
Palladium black, surface area 40-60 m2/g, 99.95% trace metals basis
Pd
205834
Palladium black, surface area 40-60 m2/g
Pd
549657
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size
O2Sn
244651
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis
O2Sn
204714
Tin(IV) oxide, ≥99.99% trace metals basis
O2Sn