EMAIL THIS PAGE TO A FRIEND
Zhonghua zhong liu za zhi [Chinese journal of oncology]
[Antitumor effect of baicalin on rat brain glioma].
PMID 23648293
Abstract
To investigate the therapeutic mechanism of baicalin on rat brain glioma. Deep brain glioma models were established by injection of glioma cell line C6 cells into the brain of Wistar rats. The rats at 7 days after modeling were randomly divided into tumor control group (0.9% NaCl solution 30 mg×kg(-1)×d(-1) gavage)and experimental groups. The experimental rats was divided into 3 groups: low dose group (50 mg×kg(-1)×d(-1)), middle dose group (100 mg×kg(-1)×d(-1)) and high dose group (200 mg×kg(-1)×d(-1)), given the baicalin by gavage. Pathological and electron microscopic changes were observed. The expressions of p53 and Bcl-2 were determined by immunohistochemistry, and the changes of MRI, the average survival time and body weight of the rats in each group after treatments were analyzed. Compared with the control group, the tumor diameter and volume of high dose group rats before sacrifice were significantly reduced (P < 0.01), and the survival time was significantly prolonged (P < 0.01). Immunohistochemistry revealed strong positive expression rate of mutant p53 (84.47 ± 3.74)% and moderately positive rate (47.28 ± 2.38)% in the control group, significantly higher than that in the negative group (12.91 ± 1.07)% (P < 0.01). The positive rate of mutant p53 of the high dose group was (46.42 ± 2.19)%, significantly lower than that of the control group (84.47 ± 3.74)% (P < 0.01). The expression rate of Bcl-2 in the control group was strongly positive (86.51 ± 4.17)% and moderate positive (48.19 ± 2.11)%, significantly higher than that of the negative group (10.36 ± 1.43)% (P < 0.01). Electron microscopy revealed that baicalin caused damages of the cell nuclei and organelles in the gliomas. Baicalin has significant inhibitory effect on glioma in vivo, and its mechanism may be related to cell apoptosis induced by down-regulated expression of mutant p53, but not related with Bcl-2 expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
572667
Baicalin, 95%
C21H18O11