EMAIL THIS PAGE TO A FRIEND
Cellular immunology
MicroRNA-155 regulates T cell proliferation through targeting GSK3β in cardiac allograft rejection in a murine transplantation model.
PMID 23648819
Abstract
Here we investigated the activity and regulation of miR-155 during cardiac allograft rejection (AR), and to examine the feasibility of using miR-155 as a biomarker of graft status. Expression of miR-155 in graft-infiltrating lymphocytes (GIL), T cells isolated from spleen (TFS), and lymphocytes separated from blood (LFB) was significantly increased during cardiac AR while GSK3β was downregulated in GIL and TFS. Inhibition of miR-155 impaired lymphocyte proliferation and enhanced the expression of GSK3β. Moreover, pharmacological inactivation of GSK3β resulted in rescue of the proliferative capability of T cells pretreated with a miR-155 inhibitor. Luciferase reporter assay confirmed that miR-155 interacted with the 3'-untranslated region (UTR) of GSK3β directly. In particular, the miR-155 in LFB can distinguish recipients with AR from syngeneic controls from POD 3 and later. The present study provides a better understanding of the pathophysiological process underlying cardiac AR progression.