EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Glucosinolate degradation products, isothiocyanates, nitriles, and thiocyanates, induce stomatal closure accompanied by peroxidase-mediated reactive oxygen species production in Arabidopsis thaliana.
PMID 23649257
Abstract
Isothiocyanates, nitriles, and thiocyanates are degradation products of glucosinolates in crucifer plants. In this study, we investigated the stomatal response to allyl isothiocyanate (AITC), 3-butenenitrile (3BN), and ethyl thiocyanate (ESCN) in Arabidopsis. AITC, 3BN, and ESCN induced stomatal closure in the wild type and the atrbohD atrbohF mutant. Stomatal closure was inhibited by catalase and salicylhydroxamic acid (SHAM). The degradation products induced extracellular reactive oxygen species (ROS) production in the rosette leaves, and intracellular ROS accumulation, NO production, and cytosolic free calcium concentration ([Ca(2+)]cyt) oscillations in guard cells, which were inhibited by SHAM. These results suggest that glucosinolate degradation products induce stomatal closure accompanied by extracellular ROS production mediated by SHAM-sensitive peroxidases, intracellular ROS accumulation, and [Ca(2+)]cyt oscillation in Arabidopsis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
308951
Ethyl thiocyanate, 97%
C3H5NS