EMAIL THIS PAGE TO A FRIEND
Nature medicine
Drug development for progeria yields insights into normal aging.
PMID 23652088
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4498
Pravastatin sodium salt hydrate, ≥98% (HPLC), powder
C23H35O7Na · xH2O