EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
Dietary liposoluble antioxidants protect mouse immune cells from the toxic effects of atmospheric ammonia.
PMID 23652442