EMAIL THIS PAGE TO A FRIEND
Journal of investigational allergology & clinical immunology
Perphenazine as a cause of mother-to-daughter contact dermatitis and photocontact dermatitis.
PMID 23653979
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6402
Perphenazine
C21H26ClN3OS