EMAIL THIS PAGE TO A FRIEND
Dentistry today
The shaping movement: fifth-generation technology.
PMID 23659098
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83440
Activated Nickel Catalyst
267015
Nickel, foil, thickness 1.0 mm, 99.995%
Ni
357553
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Ni
267007
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Ni
268259
Nickel, foil, thickness 0.125 mm, ≥99.9%
Ni
357588
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Ni
266965
Nickel, powder, <150 μm, 99.995% trace metals basis
Ni
203904
Nickel, powder, <150 μm, 99.99% trace metals basis
Ni
266981
Nickel, powder, <50 μm, 99.7% trace metals basis
Ni
268283
Nickel, powder, <1 μm, 99.8% trace metals basis
Ni
577995
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Ni
267074
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Ni
215775
Nickel, spheres, 6-16 mm, ≥99.95% trace metals basis
Ni
267058
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Ni
357626
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ni
357634
Nickel, wire, diam. 0.25 mm, ≥99.9%
Ni
767484
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ni
510033
Raney®-Nickel, W.R. Grace and Co. Raney®
2400, slurry, in H2O, active catalyst
221678
Raney®-Nickel, W.R. Grace and Co. Raney®
2800, slurry, in H2O, active catalyst
510068
Raney®-Nickel, W.R. Grace and Co. Raney®
3202, slurry, in H2O, active catalyst
510041
Raney®-Nickel, W.R. Grace and Co. Raney®
4200, slurry, in H2O, active catalyst